accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

FFFI

CAS RN 444-29-1

IUPAC Name: 1-iodo-2-(trifluoromethyl)benzene
Synonyms: 1-iodo-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 272.01
Molecular Formula: C7H4F3I
InChi Key: IGZGUYVVBABKOY-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=CC=C1I
Molecular Weight (g/mol) 
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (6)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)