accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NH2FFBr

CAS RN 444-14-4

IUPAC Name: 2-bromo-4,6-difluoroaniline
Synonyms: 2-bromo-4,6-difluoroaniline
Molecular Weight (g/mol): 208.01
Molecular Formula: C6H4BrF2N
InChi Key: WUJKFVGKLTWVSQ-UHFFFAOYSA-N
SMILES: NC1=C(F)C=C(F)C=C1Br
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results