accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3IF

CAS RN 443-85-6

IUPAC Name: 1-fluoro-3-iodo-2-methylbenzene
Synonyms: 1-fluoro-3-iodo-2-methylbenzene
Molecular Weight (g/mol): 236.03
Molecular Formula: C7H6FI
InChi Key: MSPXWJMFEVAKHQ-UHFFFAOYSA-N
SMILES: CC1=C(F)C=CC=C1I
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results