accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 443-79-8

H3C(S)CH3(S)NH2OHO

CAS RN 443-79-8

IUPAC Name: (2R,3S)-2-amino-3-methylpentanoic acid
Synonyms: Hile DL-allo-isoleucine D-alloisoleucine threo-D-isoleucine allo-D-isoleucine
Molecular Weight (g/mol): 131.18
Molecular Formula: C6H13NO2
InChi Key: AGPKZVBTJJNPAG-CRCLSJGQSA-N
SMILES: CC[C@H](C)[C@@H](N)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)