accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3PCH3H3CNNPH3CCH3CH3CH3OH3COH3COCH3OCH3CH3(?)

CAS RN 442905-33-1

IUPAC Name: 4,4'-bis[bis(3,5-dimethylphenyl)phosphanyl]-2,2',6,6'-tetramethoxy-3,3'-bipyridine
Synonyms: 4,4'-bis[bis(3,5-dimethylphenyl)phosphanyl]-2,2',6,6'-tetramethoxy-3,3'-bipyridine
Molecular Weight (g/mol): 756.86
Molecular Formula: C46H50N2O4P2
InChi Key: JRTHAKOHBMETRC-UHFFFAOYSA-N
SMILES: COC1=NC(OC)=C(C(=C1)P(C1=CC(C)=CC(C)=C1)C1=CC(C)=CC(C)=C1)C1=C(OC)N=C(OC)C=C1P(C1=CC(C)=CC(C)=C1)C1=CC(C)=CC(C)=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results