CAS RN 442633-00-3
CAS RN 442633-00-3
IUPAC Name:
2-fluoro-N-[2-(2-methyl-1H-indol-3-yl)ethyl]benzamide
Synonyms:
2-fluoro-N-[2-(2-methyl-1H-indol-3-yl)ethyl]benzamide
Molecular Weight (g/mol):
296.35
Molecular Formula:
C18H17FN2O
InChi Key:
UXRKUKRXVWJFER-UHFFFAOYSA-N
SMILES:
CC1=C(CCNC(=O)C2=CC=CC=C2F)C2=CC=CC=C2N1
1
–
2
of
2
results
1
Tocris Bioscience™ CK 666
CAS: 442633-00-3 Molecular Formula: C18H17FN2O
2
CK-666, MedChemExpress
MedChemExpress CK-666 is a cell-permeable actin-related protein Arp2/3 complex inhibitor (IC50=12 μM). CK-666 binds to Arp2/3 complex, stabilizes the inactive state of the complex, blocking movement of the Arp2 and Arp3 subunits into the activated filament-like (short pitch) conformation.