accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ClOClOClO

CAS RN 4422-95-1

IUPAC Name: benzene-1,3,5-tricarbonyl trichloride
Synonyms: benzene-1,3,5-tricarbonyl trichloride
Molecular Weight (g/mol): 265.47
Molecular Formula: C9H3Cl3O3
InChi Key: UWCPYKQBIPYOLX-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC(=CC(=C1)C(Cl)=O)C(Cl)=O
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)