accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 4412-96-8

CH3OHOO

CAS RN 4412-96-8

IUPAC Name: 3-methylfuran-2-carboxylic acid
Synonyms: 3-methylfuran-2-carboxylic acid
Molecular Weight (g/mol): 126.11
Molecular Formula: C6H6O3
InChi Key: BNYIQEFWGSXIKQ-UHFFFAOYSA-N
SMILES: CC1=C(OC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results