accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 4403-71-8

NH2NH2

CAS RN 4403-71-8

IUPAC Name: 4-(aminomethyl)aniline
Synonyms: 4-(aminomethyl)aniline
Molecular Weight (g/mol): 122.17
Molecular Formula: C7H10N2
InChi Key: BFWYZZPDZZGSLJ-UHFFFAOYSA-N
SMILES: NCC1=CC=C(N)C=C1
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results