accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 439578-83-3

NBrNHSNH2

CAS RN 439578-83-3

IUPAC Name: (6-bromopyridin-2-yl)thiourea
Synonyms: 6-bromopyridin-2-ylthiourea
Molecular Weight (g/mol): 232.1
Molecular Formula: C6H6BrN3S
InChi Key: QAIACEDQLCMXOP-UHFFFAOYSA-N
SMILES: NC(=S)NC1=NC(Br)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results