accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 4390-04-9

IUPAC Name: 2,2,4,4,6,8,8-heptamethylnonane
Synonyms: isocetane
Molecular Weight (g/mol): 226.45
Molecular Formula: C16H34
InChi Key: VCLJODPNBNEBKW-UHFFFAOYNA-N
SMILES: CC(CC(C)(C)C)CC(C)(C)CC(C)(C)C
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results