accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 4388-22-1

OSOSSOO

CAS RN 4388-22-1

IUPAC Name: {[(benzenesulfonyl)sulfanyl]sulfonyl}benzene
Synonyms: [(benzenesulfonyl)sulfanylsulfonyl]benzene
Molecular Weight (g/mol): 314.39
Molecular Formula: C12H10O4S3
InChi Key: YQUSJUJNDKUWAM-UHFFFAOYSA-N
SMILES: O=S(=O)(SS(=O)(=O)C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results