accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 4382-54-1

CH3OHNHOO

CAS RN 4382-54-1

IUPAC Name: 5-methoxy-1H-indole-2-carboxylic acid
Synonyms: 5-methoxy-1H-indole-2-carboxylic acid
Molecular Weight (g/mol): 191.19
Molecular Formula: C10H9NO3
InChi Key: YEBJVSLNUMZXRJ-UHFFFAOYSA-N
SMILES: COC1=CC=C2NC(=CC2=C1)C(O)=O
Percent Purity 
  • (1)
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)