accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 433337-11-2

CH3NNFPFFFFF

CAS RN 433337-11-2

IUPAC Name: 1-benzyl-3-methyl-1H-imidazol-3-ium; hexafluoro-λ⁵-phosphanuide
Synonyms: 3-benzyl-1-methylimidazol-1-ium hexafluorophosphate
Molecular Weight (g/mol): 318.2
Molecular Formula: C11H13F6N2P
InChi Key: VVCQOUQQPZPIKL-UHFFFAOYSA-N
SMILES: F[P-](F)(F)(F)(F)F.C[N+]1=CN(CC2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results