accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

(S)OHNOHOOOCH3CH3CH3CH3

CAS RN 4326-36-7

IUPAC Name: methyl (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoate
Synonyms: methyl (2S)-2-[(tert-butoxycarbonyl)amino]-3-(4-hydroxyphenyl)propanoate
Molecular Weight (g/mol): 295.34
Molecular Formula: C15H21NO5
InChi Key: NQIFXJSLCUJHBB-LBPRGKRZSA-N
SMILES: COC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)OC(C)(C)C
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results