accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OOHFFF

CAS RN 431-46-9

IUPAC Name: 2,2,2-trifluoro-1-methoxyethan-1-ol
Synonyms: 2,2,2-trifluoro-1-methoxyethanol
Molecular Weight (g/mol): 130.07
Molecular Formula: C3H5F3O2
InChi Key: GWTBCUWZAVMAQV-UHFFFAOYNA-N
SMILES: COC(O)C(F)(F)F
Boiling Point 
  • (3)
  • (3)
Grade 
  • (6)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (2)
  • (2)
  • (2)