accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COOOO

CAS RN 42998-51-6

IUPAC Name: 1-benzyl 3-ethyl propanedioate
Synonyms: 1-benzyl 3-ethyl propanedioate
Molecular Weight (g/mol): 222.24
Molecular Formula: C12H14O4
InChi Key: CGNOCUSLPSCMLL-UHFFFAOYSA-N
SMILES: CCOC(=O)CC(=O)OCC1=CC=CC=C1
Boiling Point 
  • (3)
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results