accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 42933-43-7

ONH2

CAS RN 42933-43-7

IUPAC Name: 2,3-dihydro-1-benzofuran-5-amine
Synonyms: 2,3-dihydro-1-benzofuran-5-amine
Molecular Weight (g/mol): 135.17
Molecular Formula: C8H9NO
InChi Key: YJMADHMYUJFMQE-UHFFFAOYSA-N
SMILES: NC1=CC2=C(OCC2)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)