accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 42732-49-0

CH3NHO

CAS RN 42732-49-0

IUPAC Name: 5-methylpyridin-3-ol
Synonyms: 5-methylpyridin-3-ol
Molecular Weight (g/mol): 109.13
Molecular Formula: C6H7NO
InChi Key: RYJNCIGFPWGVPA-UHFFFAOYSA-N
SMILES: CC1=CN=CC(O)=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)