accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ClCH3ON

CAS RN 42508-60-1

IUPAC Name: 1-(2-oxopropyl)pyridin-1-ium chloride
Synonyms: 1-(2-oxopropyl)pyridin-1-ium chloride
Molecular Weight (g/mol): 171.62
Molecular Formula: C8H10ClNO
InChi Key: JHYYWKUENNZTMD-UHFFFAOYSA-M
SMILES: [Cl-].CC(=O)C[N+]1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results