CAS RN 4238-41-9
CAS RN 4238-41-9
IUPAC Name:
hydrogen N-[(3S)-7-amino-1-chloro-2-oxoheptan-3-yl]-4-methylbenzene-1-sulfonamide chloride
Synonyms:
hydrogen C14H21clN2O3S chloride
Molecular Weight (g/mol):
369.3
Molecular Formula:
C14H22Cl2N2O3S
InChi Key:
YFCUZWYIPBUQBD-ZOWNYOTGSA-N
SMILES:
[H+].[Cl-].CC1=CC=C(C=C1)S(=O)(=O)N[C@@H](CCCCN)C(=O)CCl
1
–
1
of
1
results
1
Nalpha-(p-Toluenesulfonyl)-DL-lysine chloromethyl ketone hydrochloride, 98%
CAS: 4238-41-9 Molecular Formula: C14H22Cl2N2O3S Molecular Weight (g/mol): 369.30 MDL Number: MFCD00065395 InChI Key: YFCUZWYIPBUQBD-ZOWNYOTGSA-N Synonym: 1-chloro-3-tosylamido-7-amino-2-hepta none hydrochloride,p-toluenesulfonamide, hydrochloride 7ci, 8ci,nalpha-p-toluenesulfonyl-dl-lysine chloromethyl ketone hydrochloride PubChem CID: 23335676 IUPAC Name: N-(7-amino-1-chloro-2-oxoheptan-3-yl)-4-methylbenzenesulfonamide;hydrochloride SMILES: Cl.CC1=CC=C(C=C1)S(=O)(=O)N[C@@H](CCCCN)C(=O)CCl