accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 42333-76-6

NNNNNN

CAS RN 42333-76-6

IUPAC Name: tris(pyridin-3-yl)-1,3,5-triazine
Synonyms: tris(pyridin-3-yl)-1,3,5-triazine
Molecular Weight (g/mol): 312.34
Molecular Formula: C18H12N6
InChi Key: UOSYDSDIRAXTTF-UHFFFAOYSA-N
SMILES: C1=CC(=CN=C1)C1=NC(=NC(=N1)C1=CN=CC=C1)C1=CN=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results