accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 42204-14-8

RhCH3OO

CAS RN 42204-14-8

IUPAC Name: acetate rhodium
Synonyms: acetate rhodium
Molecular Weight (g/mol): 161.95
Molecular Formula: C2H3O2Rh
InChi Key: NENDHUHGFRLXEN-UHFFFAOYSA-M
SMILES: [Rh].CC([O-])=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results