accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OH(S)(R)O(R)OPHOHOONOHNO

CAS RN 42155-08-8

IUPAC Name: {[(2R,3S,5R)-5-(2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)-3-hydroxyoxolan-2-yl]methoxy}phosphonic acid
Synonyms: 2'-deoxyuridylic acid Dump dUMP
Molecular Weight (g/mol): 308.18
Molecular Formula: C9H13N2O8P
InChi Key: JSRLJPSBLDHEIO-SHYZEUOFSA-N
SMILES: O[C@H]1C[C@@H](O[C@@H]1COP(O)(O)=O)N1C=CC(=O)NC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results