accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 4209-91-0

CH3H3COHCH3H3C

CAS RN 4209-91-0

IUPAC Name: 3,5-dimethylhexan-3-ol
Synonyms: 3,5-dimethyl-3-hexanol
Molecular Weight (g/mol): 130.23
Molecular Formula: C8H18O
InChi Key: INMGJWCKWKKMPN-UHFFFAOYNA-N
SMILES: CCC(C)(O)CC(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results