accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 42088-91-5

H3C(S)NH3N

CAS RN 42088-91-5

IUPAC Name: (1S)-1-(pyridin-2-yl)ethan-1-aminium
Synonyms: (1S)-1-(pyridin-2-yl)ethanaminium
Molecular Weight (g/mol): 123.18
Molecular Formula: C7H11N2
InChi Key: PDNHLCRMUIGNBV-LURJTMIESA-O
SMILES: C[C@H]([NH3+])C1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results