accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NH2ICl

CAS RN 42016-93-3

IUPAC Name: 2-chloro-4-iodoaniline
Synonyms: 2-chloro-4-iodoaniline
Molecular Weight (g/mol): 253.47
Molecular Formula: C6H5ClIN
InChi Key: MYDAOWXYGPEPJT-UHFFFAOYSA-N
SMILES: NC1=CC=C(I)C=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results