accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 420137-19-5

CH3OOHN

CAS RN 420137-19-5

IUPAC Name: 4-(2-ethoxyphenoxy)piperidine
Synonyms: 4-(2-ethoxyphenoxy)piperidine
Molecular Weight (g/mol): 221.3
Molecular Formula: C13H19NO2
InChi Key: IPOFRRUDIUKVAL-UHFFFAOYSA-N
SMILES: CCOC1=CC=CC=C1OC1CCNCC1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results