CAS RN 41836-23-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Tris[N,N-bis(trimethylsilyl)amide]neodymium(III), 98%, Thermo Scientific Chemicals
CAS: 41836-23-1 Molecular Formula: C18H54N3NdSi6 Molecular Weight (g/mol): 625.403 MDL Number: MFCD03427089 InChI Key: LWKIKGQVWYVALF-UHFFFAOYSA-N Synonym: tris n,n-bis trimethylsilyl amide neodymium iii,1,1,1-trimethyl-n-trimethylsilyl silanamine neodymium iii salt,2,2,6,6-tetramethyl-n,n,3,5-tetrakis trimethylsilyl-3,5-diaza-2,6-disila-4-neodymaheptan-4-amine PubChem CID: 4397443 IUPAC Name: bis(trimethylsilyl)azanide;neodymium(3+) SMILES: C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C.[Nd+3]