accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CSiCH3H3CNSiCH3CH3H3CH3CSiCH3H3CNSiCH3CH3H3CH3CSiCH3H3CNSiCH3CH3H3CCe

CAS RN 41836-21-9

IUPAC Name: cerium(3+) tris(bis(trimethylsilyl)azanide)
Synonyms: cerium(3+) tris(bis(trimethylsilyl)azanide)
Molecular Weight (g/mol): 621.28
Molecular Formula: C18H54CeN3Si6
InChi Key: DPCZDYUESIKMSU-UHFFFAOYSA-N
SMILES: [Ce+3].C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C.C[Si](C)(C)[N-][Si](C)(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results