accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 41825-73-4

CH3CH3NH2Br

CAS RN 41825-73-4

IUPAC Name: 2-bromo-4,6-dimethylaniline
Synonyms: 2-bromo-4,6-dimethylaniline
Molecular Weight (g/mol): 200.08
Molecular Formula: C8H10BrN
InChi Key: YOSJCQJJIHEUKA-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(N)C(Br)=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)