accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 4175-77-3

IUPAC Name: 2,4-dibromo-1,3-thiazole
Synonyms: 2,4-dibromo-1,3-thiazole
Molecular Weight (g/mol): 242.92
Molecular Formula: C3HBr2NS
InChi Key: MKEJZKKVVUZXIS-UHFFFAOYSA-N
SMILES: BrC1=NC(Br)=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results