accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 41731-83-3

IUPAC Name: ethyl 2-bromo-1,3-thiazole-5-carboxylate
Synonyms: ethyl 2-bromo-1,3-thiazole-5-carboxylate
Molecular Weight (g/mol): 236.08
Molecular Formula: C6H6BrNO2S
InChi Key: KTYIFXLNIMPSKI-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN=C(Br)S1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results