accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COOOOOH3COOCH3OOCH3OOH3C

CAS RN 4163-59-1

IUPAC Name: [3,4,5,6-tetrakis(acetyloxy)oxan-2-yl]methyl acetate
Synonyms: [3,4,5,6-tetrakis(acetyloxy)oxan-2-yl]methyl acetate
Molecular Weight (g/mol): 390.34
Molecular Formula: C16H22O11
InChi Key: LPTITAGPBXDDGR-UHFFFAOYNA-N
SMILES: CC(=O)OCC1OC(OC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results