accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COHNOOCH3OCH3

CAS RN 415696-09-2

IUPAC Name: N-(3,4-dimethoxyphenyl)-2-methoxybenzamide
Synonyms: N-(3,4-dimethoxyphenyl)-2-methoxybenzamide
Molecular Weight (g/mol): 287.32
Molecular Formula: C16H17NO4
InChi Key: QLZIDCIZXPJKOS-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1C(=O)NC1=CC(OC)=C(OC)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results