accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OONaHOHHOHHOH

CAS RN 41484-91-7

IUPAC Name: sodium acetate hydrate
Synonyms: sodium acetate hydrate
Molecular Weight (g/mol): 100.05
Molecular Formula: C2H5NaO3
InChi Key: DGPIGKCOQYBCJH-UHFFFAOYSA-M
SMILES: O.[Na+].CC([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results