accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 41419-59-4

H3CNHOFFF

CAS RN 41419-59-4

IUPAC Name: N-methyl-4-(trifluoromethoxy)aniline
Synonyms: N-methyl-4-(trifluoromethoxy)aniline
Molecular Weight (g/mol): 191.15
Molecular Formula: C8H8F3NO
InChi Key: MGCCWCLGIPNIBP-UHFFFAOYSA-N
SMILES: CNC1=CC=C(OC(F)(F)F)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results