CAS RN 4136-37-2
CAS RN 4136-37-2
IUPAC Name:
hydrogen 1,1-dimethylpyrrolidin-1-ium-2-carboxylate chloride
Synonyms:
hydrogen 1,1-dimethylpyrrolidin-1-ium-2-carboxylate chloride
Molecular Weight (g/mol):
179.64
Molecular Formula:
C7H14ClNO2
InChi Key:
DUNMULOWUUIQIL-UHFFFAOYNA-N
SMILES:
[H+].[Cl-].C[N+]1(C)CCCC1C([O-])=O
1
–
1
of
1
results
1
Stachydrine hydrochloride, MedChemExpress
MedChemExpress Stachydrine hydrochloride is the major active constituent of Herba Leonuri, which is a potential therapy for cardiovascular diseases. Stachydrine can inhibit the NF-κB signal pathway. Anti-hypertrophic activities.