CAS RN 412947-54-7
CAS RN 412947-54-7
IUPAC Name:
methyl 3-amino-4-iodobenzoate
Synonyms:
methyl 3-amino-4-iodobenzoate
Molecular Weight (g/mol):
277.06
Molecular Formula:
C8H8INO2
InChi Key:
WJEBNIVVLJEIKE-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC(N)=C(I)C=C1
1
–
1
of
1
results
1
Methyl 3-amino-4-iodobenzoate, 97%
CAS: 412947-54-7 Molecular Formula: C8H8INO2 Molecular Weight (g/mol): 277.061 MDL Number: MFCD08064225 InChI Key: WJEBNIVVLJEIKE-UHFFFAOYSA-N Synonym: 3-amino-4-iodobenzoic acid methyl ester,benzoic acid, 3-amino-4-iodo-, methyl ester,methyl3-amino-4-iodobenzoate,pubchem10509,acmc-209ygt,ksc495i4l,methyl-3-amino-4-iodobenzoate,methyl 3-amino-4-iodo-benzoate,3-amino-4-iodo-benzoic acid methyl ester PubChem CID: 21628210 IUPAC Name: methyl 3-amino-4-iodobenzoate SMILES: COC(=O)C1=CC(=C(C=C1)I)N