accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 41233-93-6

KCH3CH3H3CO

CAS RN 41233-93-6

IUPAC Name: 2-methylbutan-2-ol potassium
Synonyms: 2-methyl-2-butanol potassium
Molecular Weight (g/mol): 127.25
Molecular Formula: C5H12KO
InChi Key: GGIFEFBEDYDOTI-UHFFFAOYSA-N
SMILES: [K].CCC(C)(C)O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)