accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 41230-21-1

H2NSN

CAS RN 41230-21-1

IUPAC Name: 3-(1,3-benzothiazol-2-yl)aniline
Synonyms: 3-(1,3-benzothiazol-2-yl)aniline
Molecular Weight (g/mol): 226.3
Molecular Formula: C13H10N2S
InChi Key: MKMRZZJJDIBAJF-UHFFFAOYSA-N
SMILES: NC1=CC=CC(=C1)C1=NC2=CC=CC=C2S1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)