accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 41200-96-8

IUPAC Name: 2,4-dichloro-5-(propan-2-yloxy)aniline
Synonyms: 2,4-dichloro-5-isopropoxyaniline
Molecular Weight (g/mol): 220.09
Molecular Formula: C9H11Cl2NO
InChi Key: RWRWIICNMOKBIQ-UHFFFAOYSA-N
SMILES: CC(C)OC1=CC(N)=C(Cl)C=C1Cl
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results