accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 4101-30-8

H3CONH2OCH3CH3O

CAS RN 4101-30-8

IUPAC Name: 1-(2-amino-4,5-dimethoxyphenyl)ethan-1-one
Synonyms: 1-(2-amino-4,5-dimethoxyphenyl)ethanone
Molecular Weight (g/mol): 195.22
Molecular Formula: C10H13NO3
InChi Key: KGKWXEGYKGTMAK-UHFFFAOYSA-N
SMILES: COC1=CC(N)=C(C=C1OC)C(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results