accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OOSNN

CAS RN 4100-13-4

IUPAC Name: 1,2,3-thiadiazole-4-carboxylate
Synonyms: 1,2,3-thiadiazole-4-carboxylate
Molecular Weight (g/mol): 129.11
Molecular Formula: C3HN2O2S
InChi Key: HJZYBDPHAHGHAZ-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CSN=N1
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)