accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OBOHHOOCH3

CAS RN 40972-86-9

IUPAC Name: (2,3-dimethoxyphenyl)boronic acid
Synonyms: 2,3-dimethoxyphenylboronic acid
Molecular Weight (g/mol): 181.98
Molecular Formula: C8H11BO4
InChi Key: VREWSCMOGIXMDQ-UHFFFAOYSA-N
SMILES: COC1=CC=CC(B(O)O)=C1OC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results