accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 40851-62-5

IUPAC Name: methyl 2-(2-methylphenyl)acetate
Synonyms: methyl 2-(2-methylphenyl)acetate
Molecular Weight (g/mol): 164.2
Molecular Formula: C10H12O2
InChi Key: BLEMRRXGTKTJGT-UHFFFAOYSA-N
SMILES: COC(=O)CC1=CC=CC=C1C
Percent Purity 
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results