accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 4079-68-9

H3CNCH3CH

CAS RN 4079-68-9

IUPAC Name: diethyl(prop-2-yn-1-yl)amine
Synonyms: 2-propyn-1-amine, N,N-diethyl-
Molecular Weight (g/mol): 111.19
Molecular Formula: C7H13N
InChi Key: JZJXKEWVUBVOEH-UHFFFAOYSA-N
SMILES: CCN(CC)CC#C
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)