accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H2COOOOOCH2

CAS RN 4074-88-8

IUPAC Name: 2-[2-(prop-2-enoyloxy)ethoxy]ethyl prop-2-enoate
Synonyms: diethylene glycol diacrylate
Molecular Weight (g/mol): 214.22
Molecular Formula: C10H14O5
InChi Key: LEJBBGNFPAFPKQ-UHFFFAOYSA-N
SMILES: C=CC(=O)OCCOCCOC(=O)C=C
Boiling Point 
  • (2)
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results