accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

FFFOBr

CAS RN 407-14-7

IUPAC Name: 1-bromo-4-(trifluoromethoxy)benzene
Synonyms: 1-bromo-4-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 241.01
Molecular Formula: C7H4BrF3O
InChi Key: SEAOBYFQWJFORM-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=CC=C(Br)C=C1
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)