accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 4052-30-6

IUPAC Name: 4-methanesulfonylbenzoic acid
Synonyms: P-methylsulfonylbenzoic acid
Molecular Weight (g/mol): 200.21
Molecular Formula: C8H8O4S
InChi Key: AJBWNNKDUMXZLM-UHFFFAOYSA-N
SMILES: CS(=O)(=O)C1=CC=C(C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results